Principaux documents
EP022639200
Eppendorf® Adapter
to hold, 2 x 15mL, for 100 mL round bucket in Rotor A-4-38, pack of 2 ea
About This Item
Code UNSPSC :
41103911
Matériaux
POM
Caractéristiques
to hold 2 x 15mL, for 100 mL round bucket in Rotor A-4-38
Conditionnement
pack of 2 ea
Fabricant/nom de marque
Eppendorf® 022639200
Compatibilité
Rotor A-4-38
Catégories apparentées
Description générale
Adapter, for 2 conical tubes 15 mL, 17.2 mm × 121 mm, for 100 mL round bucket in Rotor A-4-38, 2 pcs.
  • for 100 mL round bucket in Rotor A-4-38
  • Please note that this adapter is not compatible with the aerosol-tight caps of this rotor.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG