EP0030128591
Eppendorf® Twin tec PCR plates, 96 wells
green plate, conical bottom colorless wells, semi-skirted, pkg of 25 ea
154,00 €
About This Item
154,00 €
Produits recommandés
Matériaux
conical bottom colorless wells
green plate
polycarbonate
Stérilité
non-sterile
Caractéristiques
skirt (Semi skirted)
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Eppendorf® 0030128591
Technique(s)
PCR: suitable
Taille
96 wells
Volume de travail
250 μL
Description générale
Caractéristiques et avantages
- Eppendorf® Twin tec PCR plate combine a polycarbonate frame and polypropylene wells for optimum performance.
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
- Exceptionally solid and torque-resistant polycarbonate frame.
- Cut-away corner and alphanumeric grid-referencing.
- Raised well rims for effective sealing, also reduces risk of cross-contamination.
- Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors.
- Fits most thermal cyclers.
- 250 μL max. well volume (when used with cap strips).
- Improved well-to-well tolerance.
- Ideal for quantitative real-time PCR as well – OptiTrack® matrix for faster sample identification and fewer pipetting errors.