Principaux documents
EP0030108450
Eppendorf® Protein LoBind Tubes
PCR clean, withstand 18,000 x g, 2.0 mL, pack of 500 ea
Synonyme(s) :
Eppendorf® Protein LoBind tubes
500 EA
89,40 €
89,40 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
500 EA
89,40 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.15
89,40 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
colorless
conical bottom
Description
Low retention
Stérilité
non-sterile
Caractéristiques
DNase free
PCR clean
Conditionnement
pack of 500 ea
Fabricant/nom de marque
Eppendorf® 0030108450
Capacité
2.0 mL
Adéquation
suitable for PCR
Description générale
Avoid losing valuable samples during centrifugation, incubation and storage! Rely on the excellent protection offered by the hinged Safe-Lock lid. It contains all of our experience from 50 years of continuous optimization and development. Trust the original Eppendorf Safe-Lock Tubes, because your samples deserve only the best.
Caractéristiques et avantages
Eppendorf Protein LoBind Tubes are specifically designed to ensure you get maximal protein recovery.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG