Principaux documents
EP0030076257
Eppendorf® Empty Box 2.0 for epT.I.P.S.® Reloads
for tips 1000 – 1,250 µL
About This Item
Code UNSPSC :
41121800
Caractéristiques
for tips 1000 – 1,250 µL
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Eppendorf® 0030076257
Volume
1000—1250 μL
Catégories apparentées
Description générale
epT.I.P.S.® Box 2.0 for epT.I.P.S.® Reloads, empty box, does not include tips, Eppendorf Quality, 1,000 – 1,250 µL
Application
General liquid handling
Caractéristiques et avantages
  • epT.I.P.S.® Box 2.0 – empty reusable boxes for tip volumes of 10 µL - 5 mL
  • epT.I.P.S.® Box 2.0 – all three sizes are availabe in Eppendorf Quality
  • Autoclavable at 121 °C (20 min.) for subsequent use
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG