Principaux documents
DWK261352451
DURAN® culture tubes
new
GL 18 thread, tube capacity 34 mL, with cap, borosilicate glass 3.3 tube
About This Item
Code UNSPSC :
41121706
Matériaux
PBT cap
PTFE seal
borosilicate glass 3.3 tube
Joint
joint threaded neck
Caractéristiques
autoclavable
with cap
Conditionnement
pack of 50 ea
Fabricant/nom de marque
DWK Life Sciences DURAN® 261352451
glass tube diam. × H
20 mm × 150 mm
Capacité du tube
34 mL
Raccords
GL 18 thread
Adéquation
suitable for (culture)
Description générale
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today′′s research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used. The original DURAN® culture tubes are distinguished by their uniform wall thickness distribution, minimal thermal expansion and the resulting high resistance to temperature changes. They are therefore particularly suitable for work at very high temperatures and rapid temperature changes.
Application
In vitro cell culture
Caractéristiques et avantages
  • Manufactured from borosilicate glass 3.3
  • With DIN thread and screw cap from PBT for airtight sealing
  • High tightness due to PTFE-coated silicone seal
Informations légales
DURAN is a registered trademark of DWK Life Sciences