Documents
CLS6770-RTA20
Corning® LSE Adapter
0.20 for rotor (Cat. No. 6770-RT)
Synonyme(s) :
Corning® LSE Adapters
About This Item
Code UNSPSC :
23152104
Nomenclature NACRES :
SB.56
Caractéristiques
0.20 for rotor (Cat. No. 6770-RT)
Fabricant/nom de marque
Corning 6770-RTA.20
Adéquation
suitable for (for use with Corning LSE compact centrifuge)
Description générale
These are optional adapters for Corning LSE compact centrifuge rotors: CLS480136, CLS480137, CLS480138; CLS480139
Informations légales
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.