Principaux documents
CLS3121150CO
Pyrex® small top desiccators with 24/29 standard taper stopcock
replacement cover for 2.4L large top desiccator w/ 24/29 standard taper stopcock, 1/cs
Synonyme(s) :
glass desiccator cover, glass desiccator lid
About This Item
Code UNSPSC :
41104510
Nomenclature NACRES :
NB.43
Le produit CLS3121150CO n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Matériaux
borosilicate glass (borosilicate glass code 7740)
Caractéristiques
reusable
Conditionnement
case of 1
pack of 1
Fabricant/nom de marque
Corning 3121-150CO
Hauteur
85 mm
Description générale
These PYREX desiccator covers are replacements for 2.4L desiccators CLS3121150. They require a replacement 24/29 Standard Taper stopcock with a tooled hose connection. The plug material is PTFE.
Informations légales
PYREX is a registered trademark of Corning, Inc.