Principaux documents
CLS11650
Corning® closed system bottle
capacity 150 mL, polystyrene, male Luer Lock, sterile
Synonyme(s) :
Closed System Bottle, Corning System Bottle
8 EA
2 050,00 €
2 050,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
8 EA
2 050,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
2 050,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
clear bottle
high-density polyethylene cap
polystyrene
round bottle
wide-mouth bottle
Stérilité
sterile
Caractéristiques
cap
male Luer Lock
Conditionnement
pkg of 8 ea
Fabricant/nom de marque
Corning 11650
Diam. du col du flacon
~42 mm
Capacité
150 mL
Description générale
Corning polystyrene Storage Bottles are now available as closed system solutions. They are sterile and arrive individually double-bagged and preassembled.
The preassembled transfer caps have two ports.
One port ends in a 0.2 μm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in a male Luer Lock. The dip tube reaches all the way to the bottom of the flask for easy aseptic transfer of your liquid handling processes.
  • Made from optically clear polystyrene
  • Sterilized to SAL 10-6
Informations légales
Corning is a registered trademark of Corning, Inc.