Principaux documents
FW 11.9.2.2
Reactivity: Anti-CD44 v6, human, 93070778
About This Item
Code UNSPSC :
41106514
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Source biologique
Mouse x Mouse Hybridoma
ImmunoDon
BALB/c mouse spleen
Myélome
Sp2/0
Réactivité
Anti-CD44 v6, human
Technique(s)
cell culture | mammalian: suitable
Isotype
IgG2a
Conditions d'expédition
dry ice
Température de stockage
−196°C
Origine de la lignée cellulaire
Mouse BALB/c x Mouse Sp2/0 myeloma
Description de la lignée cellulaire
The antibody is directed against the variant region of human CD44 encoded by exon 6. The antibody is suitable for immunohistology, but if used for FACS staining binding must be performed at room temperature.
Immunogène
Fusion protein of human CD44 v6
Application
Immunohistology and FACS
Milieu de culture
IDMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Procédure de repiquage
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Autres remarques
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.