C1046
Silicone sponge closures
size 38 mm
Synonyme(s) :
Closures for Vials, Silicone Closures, Sponge Closures
About This Item
Produits recommandés
Taille
38 mm
Catégories apparentées
Description générale
Silicone rubber rings with sealed-in silicone sponge filters.
These silicone sponge closures are ideal when working with aerobic cultures; they permit an efficient exchange of gases. They also cut losses due to evaporation in half in comparison to cotton plugs, thus minimizing changes in pH values. They can be dry heat sterilized repeatedly at 180°C and are easy to clean and maintain (reusable).
These silicone sponge closures are ideal when working with aerobic cultures; they permit an efficient exchange of gases. They also cut losses due to evaporation in half in comparison to cotton plugs, thus minimizing changes in pH values. They can be dry heat sterilized repeatedly at 180°C and are easy to clean and maintain (reusable).
Compatibilité
For use with 500-1000 mL Erlenmeyer and shake flasks.