BMSIPD960015ZOB
BeadBlaster™ 96 Zirconium Oxide Grinding Ball
bead diam. 15 mm, Zirconium Oxide Grinding grinding jar
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NA.21
Produits recommandés
Matériaux
(Zirconium Oxide Grinding)
Conditionnement
pack of 2
Fabricant/nom de marque
Benchmark Scientific IPD9600-15ZOB
Diam. des billes
15 mm
Compatibilité
Zirconium Oxide Grinding grinding jar
Description générale
Zirconium Oxide is the most durable and efficient media for ball milling and attrition milling of ceramic materials. Extremely low abrasive wear prevents materials from being polluted. Suitable for the environment of high viscosity, wet grinding and dispersion. It shows high grinding efficiency and long-time working life. Zirconia grinding media provides a virtually contamination-free ball milling solution for a variety of industries.
Application
Homogenization of metallurgy, geology and pharmaceutical samples
Caractéristiques et avantages
- virtually contamination
- free ball milling
- High hardness for wear resistance
- Not corroded by exposure to solvents
Informations légales
BeadBlaster is a trademark of Benchmark Scientific, Inc.