Principaux documents
BMSC1005T5S
Sigma-Aldrich
Microtube Centrifuge Tubes
capacity 5 mL, clear tube, sterile, cap (attached pushed fit), pack of 20 ea (10 x bags 20ea)
About This Item
Code UNSPSC :
41121703
Matériaux
cap (attached pushed fit)
clear tube
conical bottom
polypropylene tube
Stérilité
sterile
Caractéristiques
DNase free
RNase free
graduations
Conditionnement
pack of 20 ea (10 x bags 20ea)
Fabricant/nom de marque
Benchmark Scientific C1005-T5-S
Capacité
5 mL
Diam. du tube
16 mm
Adéquation
suitable for (c
entrifugation)
suitable for PCR
Description générale
5mL MacroTubes take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption. Useful from -86°C to 110°C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from gamma sterilized racked packaging with attached caps or non-sterile bulk packaging with separate caps. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space.
Application
Centrifugation, PCR