Principaux documents
BAH370800020
Bel-Art® Vortexer Attachment for Microcentrifuge Tubes
for tubes, 1.5 mL, no. of holes, 20, polypropylene
Synonyme(s) :
tube holder
About This Item
Code UNSPSC :
41121702
Nomenclature NACRES :
NB.22
Matériaux
polypropylene
Caractéristiques
for tubes 1.5 mL
no. of holes 20
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art H37080-0020
Description générale
Simple to use, hold the shaft end against the rubber pad or test tube cup of a vortexer and quickly mix eight or twenty 1.5ml tubes. Rugged polypropylene construction. 165mm (6 1/2") long shaft has a 25.4mm (1") diameter ball handle. 1 each,
Simultaneously vortex multiple 1.5ml microcentrifuge tubes using a standard vortexer.
  • Two capacities available, 8 and 20 tube
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP