AXYVT3845ULR
Pipette tips in 384-tip format, robotic
size 5 μL, pkg of 5x3840tips/cs, racked
Synonyme(s) :
pipet tips
4 690,00 €
About This Item
Corning
4 690,00 €
Fabricant/nom de marque
Axygen Inc VT-384-5UL-R
Corning
Conditionnement
pkg of 5x3840tips/cs
Taille
5 μL
Description générale
- Axygen® automation pipette tips in 384-tip format are designed for Agilent®/Velocity11 VPrep® and Bravo 96LT, 8LT, 384ST, 96ST, and 16ST head.
- Manufactured to stringent specifications including automated assembly to minimize contamination risks and ensure consistent performance and quality.
- The clear polypropylene high-quality tips are ideal for sensitive biological applications.
- RNase-/DNase-free and nonpyrogenic.