Principaux documents
ADF356309-EP
Adapter for Eppendorf® F-35-6-30 Rotor
holds 1 x 9 - 15 mL round-bottom tube and blood collection tube, large rotor bore, large rotor bore, pack of 2 ea
About This Item
Code UNSPSC :
41103911
Caractéristiques
holds 1 x 9 - 15 mL round-bottom tube and blood collection tube, large rotor bore
Conditionnement
pack of 2 ea
Fabricant/nom de marque
Eppendorf® 022654545 (NA)
Paramètres
7,403 × g max. RCF
Technique(s)
centrifugation: suitable
Diamètre
17 mm
Compatibilité
for use with Rotor F-35-6-30 (large rotor bore)
Description générale
Adapters to fit various size tubes and bottles for use with Eppendorf rotor F-35-6-30.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG