Principaux documents
RF09NC
BS/IP/RF U-Tube Reverse Flow Viscometer for Transparent and Opaque Liquids
(Size 9, Nom.Constant 30, Range 6000 to 30,000 cSt)
About This Item
Code UNSPSC :
41116107
Nomenclature NACRES :
NA.24
Fabricant/nom de marque
Paragon Scientific Ltd
Application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care