Principaux documents
32438
Supelco
Parbendazole
VETRANAL®, analytical standard
Synonyme(s) :
Methyl (5-butyl-1H-benzoimidazol-2-yl)carbamate, Methyl 5(6)-butyl-2-benzimidazolecarbamate
25 MG
77,50 €
77,50 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Changer de vue
25 MG
77,50 €
About This Item
Formule empirique (notation de Hill) :
C13H17N3O2
Numéro CAS:
Poids moléculaire :
247.29
Numéro CE :
Numéro MDL:
Code UNSPSC :
41116107
ID de substance PubChem :
Nomenclature NACRES :
NA.24
77,50 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Qualité
analytical standard
Niveau de qualité
Gamme de produits
VETRANAL®
Durée de conservation
limited shelf life, expiry date on the label
Technique(s)
HPLC: suitable
gas chromatography (GC): suitable
Application(s)
forensics and toxicology
pharmaceutical (small molecule)
Format
neat
Température de stockage
2-8°C
Chaîne SMILES 
CCCCc1ccc2nc(NC(=O)OC)[nH]c2c1
InChI
1S/C13H17N3O2/c1-3-4-5-9-6-7-10-11(8-9)15-12(14-10)16-13(17)18-2/h6-8H,3-5H2,1-2H3,(H2,14,15,16,17)
Clé InChI
YRWLZFXJFBZBEY-UHFFFAOYSA-N
Description générale
Parbendazole is a carbamate ester-amine. It finds applications in control or treatment of nematode infestations.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Informations légales
VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Pictogrammes
Health hazardExclamation mark
Mention d'avertissement
Warning
Mentions de danger
Classification des risques
Acute Tox. 4 Oral - Repr. 2
Code de la classe de stockage
11 - Combustible Solids
Classe de danger pour l'eau (WGK)
WGK 3
Hiroshi Sato et al.
The Journal of veterinary medical science, 65(4), 453-457 (2003-05-09)
Larva migrans caused by the common raccoon ascarid, Baylisascaris procyonis, is a zoonotic disease of critical importance in North America. Recently we encountered the first proven outbreak of this disease in Japan in domestic rabbits (Oryctolagus cuniculus) in a small
N C Sangster et al.
The Journal of parasitology, 71(5), 645-651 (1985-10-01)
Benzimidazole treatment produced greater effects on microtubule-dependent acetylcholinesterase secretion, the presence of microtubules in intestinal cells, and colchicine binding in susceptible compared with benzimidazole-resistant Trichostrongylus colubriformis. In addition, the binding of benzimidazoles was markedly reduced in preparations from the latter
K E Foster et al.
European journal of biochemistry, 163(3), 449-455 (1987-03-16)
The mutant BEN210 of Physarum polycephalum is highly resistant to a number of benzimidazole carbamate agents, including methylbenzimidazole-2-yl-carbamate and parbendazole. The resistance is conferred by the benD210 mutation in a structural gene for beta-tubulin. This mutant allele encodes a beta-tubulin
E Lacey et al.
Molecular and biochemical parasitology, 19(2), 171-181 (1986-05-01)
The binding of tritiated benzimidazoles (BZs)-albendazole, parbendazole, oxibendazole, mebendazole, oxfendazole and fenbendazole-to crude tubulin extracts from BZ-susceptible and -resistant Haemonchus contortus has been examined. For all BZs, the binding was substantially lower in the resistant isolate. The extent of this
D R Hennessy et al.
Journal of veterinary pharmacology and therapeutics, 15(1), 10-18 (1992-03-01)
The effect of intraruminal administration of parbendazole (PBZ) on the flow rate of bile and the pharmacokinetic behaviour of oxfendazole (OFZ) was examined in sheep. PBZ given at 18, 9 and 4.5 mg/kg resulted in a dose-related reduction in bile