Principaux documents
37-0090
Sigma-Aldrich
Zinc
JIS special grade
About This Item
Formule empirique (notation de Hill):
Zn
Numéro CAS:
Poids moléculaire :
65.39
Numéro MDL:
Code UNSPSC :
12141750
ID de substance PubChem :
Qualité:
JIS special grade
Forme:
granular
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Qualité
JIS special grade
Pression de vapeur
1 mmHg ( 487 °C)
Forme
granular
Pertinence de la réaction
reagent type: catalyst
core: zinc
Disponibilité
available only in Japan
Résistivité
5.8 μΩ-cm, 20°C
pb
907 °C (lit.)
Pf
420 °C (lit.)
Densité
7.133 g/mL at 25 °C (lit.)
Température de stockage
15-25°C
Chaîne SMILES 
[Zn]
InChI
1S/Zn
Clé InChI
HCHKCACWOHOZIP-UHFFFAOYSA-N
Code de la classe de stockage
13 - Non Combustible Solids
Classe de danger pour l'eau (WGK)
WGK 3
Point d'éclair (°F)
Not applicable
Point d'éclair (°C)
Not applicable
Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
Casper Groth et al.
Development (Cambridge, England), 140(14), 3018-3027 (2013-06-21)
Developmental patterning requires the precise interplay of numerous intercellular signaling pathways to ensure that cells are properly specified during tissue formation and organogenesis. The spatiotemporal function of the Notch signaling pathway is strongly influenced by the biosynthesis and intracellular trafficking
Bikha Ram Devrajani et al.
JPMA. The Journal of the Pakistan Medical Association, 63(3), 369-373 (2013-08-07)
To determine the serum zinc level in patients with pneumonia. The descriptive cross-sectional study was conducted at the Liaquat University Hospital, Hyderabad, Pakistan, from September 2008 to February 2009. All patients above 12 years of age and of either gender
Lena Martin et al.
Archives of animal nutrition, 67(3), 248-261 (2013-06-08)
This study aimed at investigating the effect of dietary zinc on performance, jejunal brush border membrane enzyme activities and mRNA levels of enzymes and two zinc transporters in piglets. A total of 126 piglets were weaned at 26 ±1 days
Michael Alvares et al.
Pediatrics, 132(1), e229-e232 (2013-06-05)
As food allergies become increasingly prevalent and testing methods to identify "food allergy" increase in number, the importance of careful diagnosis has become even more critical. Misdiagnosis of food allergy and inappropriate use of unproven testing modalities may lead to