Principaux documents
MXPPA4RP1
Millipore
Milliflex® Plus Essential Requalification Protocol
A4 format
About This Item
Code UNSPSC :
41116500
Nomenclature NACRES :
NB.79
Compatibilité
for use with Milliflex® Plus pump.
Description générale
Milliflex® Plus Essential Requalification protocol is designed as a ready to use guide to perform the requalification of the Milliflex® Plus pump. The requalification protocol can be used to re-certify consistent performance of the pumps upon encountering any change or at regular intervals.
Application
Milliflex® Plus Essential Requalification protocol is used for the re-qualification of Milliflex® Plus pump which is used to perform bioburden testing for pharmaceutical products.
Informations légales
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany