Principaux documents
1.16983
Supelco
Iron Test
About This Item
Code UNSPSC :
41116116
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Niveau de qualité
Description générale
The Reflectoquant® test strips offer a very simple means of determining iron(II) in surface water, seawater, industrial water, wastewater and mineral water. Furthermore samples of food and preserves can be analyzed. They can thus be used to distinguish between ferrous and non-ferrous metals. On the test strip, in acidic solution iron(II) ions react with 2,2′-bipyridine to form a red complex that is determined reflectometrically on the RQflex®. For a determination of Fe(III) it is necessary to treat the sample with Ascorbic acid first, to reduce Fe(III) to Fe(II). For pre-testing a sample in an economic and quick way to estimate the iron concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant® test strips for iron. Further quantitative determination methods can be found with our Spectroquant® tests.
Informations légales
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany