Principaux documents
Z765376
Microflex® Ultraform® nitrile gloves
size L
Synonyme(s) :
disposable glove
About This Item
Code UNSPSC :
46181504
Nomenclature NACRES :
NB.35
Le produit Z765376 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Matériaux
blue nitrile
Taille
L
Stérilité
non-sterile
non-sterile
Caractéristiques
CE compliant: no
disposable
latex free
powder-free
durability, standard weight
powder free, textured tips, no lining and not water resistance
Conditionnement
case of 3000 ea (10 x 300e)
pack of 300 × gloves
Fabricant/nom de marque
Microflex UF-524-L
L × épaisseur
230 mm × 0.05 mm , size type, ambidexterous
Description générale
The unique soft formulation of Ultraform® fits your hand effortlessly as if it is a second skin. In field tests users wearing Ultraform actually experienced an increase in fine motor task performance compared to a bare hand.
You have to feel how this feels for yourself.
Informations légales
Microflex is a registered trademark of CIT Healthcare LLC
Ultraform is a registered trademark of CIT Healthcare LLC