This item is not tinted or intended for use as a UV shielding device. The polycarbonate shield offers protection in chemical splash events, cutting or grinding, spraying, or wood and metal work.
Z759015
Bollé Sphere safety visor with headgear & brow guard
CE compliant
Synonyme(s) :
protective eyewear, protective glasses, safety eyeglasses, safety eyewear, safety glasses
31,50 €
About This Item
31,50 €
Produits recommandés
Caractéristiques
CE compliant
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Bollé SPHERE
Disponibilité
not available in USA
Description générale
- Ultra enveloping face shield
- Adjustable headgear and brow guard
- Polycarbonate visor
-
Does the following visor Z759015 Bollé Sphere safety visor with headgear & brow guard protect from the UV light?