Principaux documents
Z742311
Accuris E2 Grade Calibration Weight Set
1 EA
709,00 €
709,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
709,00 €
About This Item
Code UNSPSC :
41111500
Nomenclature NACRES :
NB.71
709,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Fabricant/nom de marque
Benchmark Scientific W1101-6-100
Description générale
Premium Class E2 Weight Set is ideal for calibration of analytical balances with a 0.0001g readability. Precision manufactured from high purity, 316 stainless steel, these weights are corrosion resistant with magnetic susceptibility <0.005. The set includes 6 weights, 100mg to 100g, for checking repeatability, cornerload, linearity and span. The weights are supplied in a protective case with tweezers, a cleaning cloth and a certificate of verification.
Conditionnement
6 pcs: 1x100 mg, 1 x 10 g, 2 x 20 g, 1 x 50 g, 1 x 100 g, includes protective case, tweezers, dust brush and cleaning cloth
Informations légales
Accuris is a trademark of Benchmark Scientific, Inc.