Principaux documents
Z740967
Sigma-Aldrich
Aldrich® Essentials culture tube with screw cap and rubber liner
capacity 50 mL, O.D. × L 25 mm × 150 mm
About This Item
Code UNSPSC :
41121706
Nomenclature NACRES :
NB.11
Matériaux
borosilicate glass tube
round bottom
screw top cap
screw top neck
transparent tube
Caractéristiques
neck thread external thread
Conditionnement
pack of 192 ea
Fabricant/nom de marque
Sigma-Aldrich
D.E. × L
25 mm × 150 mm
Capacité
50 mL
Description générale
Clear borosilicate glass, round bottom culture tubes with thread top and black phenolic cap. Cap has plain rubber liner. Conforms to ASTM E982.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC