Z682144
Ace PTFE lid only for condenser "C" and "CR" assemblies
fits Büchi® models R-200/R-205 and R-114/R-144
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.43
Produits recommandés
Fabricant/nom de marque
Büchi equivalent, 27479
Description générale
The PTFE lid fits the Ace condenser "C" and "CR" assemblies.
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG