Z677957
Finntip® Flex pipette tips
Finntip Flex 300, non-sterile
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Produits recommandés
Matériaux
colorless tip
orange
Stérilité
non-sterile
Caractéristiques
barrier: no
Conditionnement
pack of 960 ea (Racks)
Fabricant/nom de marque
Finn 94060510
Volume maximum
300 μL
Gamme de volumes
5-300 μL
Description générale
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
Autres remarques
Browse all pipette tips
Informations légales
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries