Z677531
Lapren™ latex/chloroprene flock lined gloves
size S
Synonyme(s) :
chemically resistant glove
353,00 €
About This Item
353,00 €
Produits recommandés
Matériaux
green (chloroprene, natural latex)
green gloves
Taille
S
Stérilité
non-sterile
Caractéristiques
CE compliant
disposable
latex free: no
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
Conditionnement
package of 100 ea
Fabricant/nom de marque
KCL 706 - 07
KCL 706
L × épaisseur
310 mm × 0.6 mm
Description générale
With cotton flock lining, stretched cuff and anatomical design, this product meets all requirements of comfort and hygiene.
- Ergonomic fit and high elasticity ensures good sensitivity
- Profiled palm for firm grip when wet
- High mechanical resistance
- Cotton flock lining improves comfort
- Free from allergenic substances such as thiurames, benzothiazoles and thiourea
- CE compliant