Z677302
Dermatril™ nitrile gloves
size XXL
Synonyme(s) :
Disposable Nitrile Gloves, Nitrile Laboratory Gloves, disposable glove
481,00 €
About This Item
481,00 €
Produits recommandés
Matériaux
blue gloves
blue nitrile
Taille
XXL
Stérilité
non-sterile
Caractéristiques
CE compliant
latex free
powder-free
Conditionnement
pack of 100 ea (boxes of 100)
Fabricant/nom de marque
KCL 740 - 11
KCL 740
L × épaisseur
250 mm × 0.11 mm
Description générale
All Dermatril disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
Caractéristiques et avantages
- Anti-static.
- Free from allergenic substances such as thiurames, benzothiazoles and thiourea.
- Roughened fingertips for better grip.
- Rolled cuff edge for easier donning.
- CE compliant.
- Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 455, EN 61340-5-1, ASTM 1671:2001