Principaux documents
Z665746
Ohaus® Discoverysemi-micro analytical balances
DV314C, AC/DC input 230 V AC
Synonyme(s) :
analytical scales, laboratory balances, weighing balances
About This Item
Code UNSPSC :
41111502
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
stainless steel weighing plate
Caractéristiques
CE compliant
CSA approved
Fabricant/nom de marque
DV314C
Plage de mesure de la capacité de pesée
310 g, resolution: 0.2 mg
Diam. de la plateforme
90 mm
Description générale
Ohaus Discovery series of semi-micro and analytical balances combine unmatched weighing performance with Ohaus′ innovative SmarText software, making it extremely reliable and easy-to-use. The Discovery features durable glass and steel construction and together with advanced AutoCal internal calibration makes it the best performing balance in its class.
Discovery models have capacities upto 310g and are perfect for university research, pharmaceutical research and drug discovery, and general lab applications.
Caractéristiques et avantages
  • Highest performance
  • Multiple weighing units
  • Environmental settings
  • Easy to set up and use
  • Advanced AutoCal calibration
  • Four methods of density determination
  • RRS232 interface with full GLP/GMP Protocol
  • Protective in-use cover
  • Up-front level indicator
Informations légales
Ohaus is a registered trademark of Ohaus Corp.
Les clients ont également consulté
Slide 1 of 1