Principaux documents
Z660604
Plas-Labs temperature and humidity control box
AC/DC input 240 V AC (UK plug)
Synonyme(s) :
isolation chambers, pcr chambers
1 EA
34 970,00 €
34 970,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
34 970,00 €
About This Item
Code UNSPSC :
41103406
Nomenclature NACRES :
NB.35
34 970,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Fabricant/nom de marque
Plas-Labs 890-THC/DT/EXP/UK
Intérieur, Haut. × larg. × Prof.
26 in. × 41 in. × 28 in.
Extérieur, Haut. × larg. × Prof.
38 in. × 55 in. × 35 in.
Description générale
Price includes shipping
The Plas-Labs temperature and humidity control glove box is designed to maintain accurate control of the isolated atmosphere. The drying train can be used when in-house inert gas is not available. The molecular sieve should be regenerated periodically for optimal drying performance.
The easy to use touch screen offers password protection, on screen readings of chamber temperature and humidity, full color screen, and room temperature and humidity readings.
  • Humidity (Rh) transmitter with 8ft. sensor cable
  • Two inlet/outlet gas valves on main chamber and transfer chamber
  • Serial port connection for data logging
  • Thermoelectric assembly (Peltier device)
  • Nebulizer
  • Two drying train canisters (filled with Molecular Sieve) and circulation pump
  • CE compliant