Principaux documents
Z656097
Heidolph Reax 2 overhead shaker
AC/DC input 230 V AC
1 EA
1 240,00 €
1 EA
1 240,00 €
1 EA
1 620,00 €
1 240,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
1 240,00 €
1 EA
1 240,00 €
1 EA
1 620,00 €
About This Item
Code UNSPSC :
41103811
Nomenclature NACRES :
NB.73
1 240,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
CE compliant
Fabricant/nom de marque
541-21009-00 (EU)
541-21009-03 (UK)
541-21009-04 (CH)
Disponibilité
available only in EU
Paramètres
20-100 rpm speed
Description générale
  • Superior in hematology, medical diagnostics, materials testing and biochemistry.
  • Comes with Easy-to-change universal adapter for use of various vessels
  • Adapter for use of up to 20 test tubes at the same time (accessory)
  • Comes with non-skid rubber mat
  • Analogue rpm adjustable dial controls
  • Protection class IP 21
Questions
  1. Does this include universal adapter? Also, please provide me full description of the product.
    1 answer
    1. Yes, this product includes a universal adapter. Additional specifications are listed below: