Principaux documents
Z646318
Sigma-Aldrich
Aldrich® drum fittings
vented flex spout for 60 L Aldrich drums
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.51
Diamètre
60 mm
Description générale
Robust PP drum fittings designed to fit Aldrich 5, 10, 20, 30 & 60 L plastic drums, making chemical dispensing from our containers safe and simple.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC