Z630039
IKA® HS 501 Horizontal shaker
AS501.1 Universal attachment
Synonyme(s) :
IKA® HS 501 horizontal shaker universal attachment, shaking plate
Conditionnement
pack of 1 ea
Fabricant/nom de marque
IKA 0008000200
Larg. × P × H
480 mm × 500 mm × 120 mm
Description générale
Using the universal attachment, all kinds of vessels can be adapted by universal, infinitely variable clamping rolls
Informations légales
IKA is a registered trademark of IKA-Werke GmbH & Co. KG