Principaux documents
Z561894
Sigma-Aldrich
Aldrich® glass beaker with handle
capacity 300 mL, borosilicate glass
Synonyme(s) :
beaker mug
About This Item
Code UNSPSC :
41121803
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
Caractéristiques
graduations
Fabricant/nom de marque
Sigma-Aldrich
Capacité
300 mL
Description générale
Made from 100% borosilicate glass with white, easy to read, graduations on back. With a 300 mL capacity, the large side handle stays cool when hot liquids are inside. These beaker mugs are heatproof, ovenproof, microwave and dishwasher safe.
The attached handle eliminates the need for clumsy gripping devices when handling hot liquids. Beakers are made of lightweight glass that is durable and has excellent heat transfer properties for rapid heating and cooling.
Autres remarques
Browse all glass beakers
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC