Z551511
Torion™ screw threaded plastic plug
#14
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Produits recommandés
Description
For 14 mm tubing
Conditionnement
pkg of 5 ea
Description générale
#8 will fit a 5/16in. O.D. size, and #14 will fit a 1/2in. O.D. size, and 3/16in. O.D. fits a #5 Torion.
Informations légales
Torion is a trademark of Bibby Sterlin Ltd.