Documents
Z532266
Sigma-Aldrich
Aldrich® pear-shaped flask
capacity 5 mL, joint: ST/NS 14/20
Synonyme(s) :
pear flask, pear shape flask, round flasks
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
pear-shaped flask
short
single-neck
Joint
joint: ST/NS14/20
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Sigma-Aldrich
Capacité
5 mL
Description générale
Short neck.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC