This product is an Acetal plastic, also known as polyoxymethylene (POM).
Z503371
Scienceware® clip clamp
midi
18,40 €
About This Item
18,40 €
Produits recommandés
Caractéristiques
for tubing o.d. 3.2 ‑ 11 mm
Conditionnement
pkg of 12 ea
Fabricant/nom de marque
Bel-Art H18228-0000
-
What kind of plastic is this product made out of?