Principaux documents
Z424595
Pressure relief valve for PLAS-LABS glovebox
1 EA
363,00 €
363,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
363,00 €
About This Item
Code UNSPSC :
42132201
Nomenclature NACRES :
NB.35
363,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Fabricant/nom de marque
Plas-Labs 800-PRV
Description générale
Useful for eliminating unwanted positive pressure build up in any of Plas-Labs® glove boxes. The acrylic relief valve is easily mounted at the gas inlet or outlet with 1/8 in. NPT SS connections. Exhausted air can be directed away from operators by a hose connection to the outlet port. For minimizing gas consumption, use with the controlled purge system.