Documents
Z309265
Quickfit® flat-bottom boiling flasks
capacity 50 mL, joint: ST/NS 19/26
Synonyme(s) :
FF50/2S
About This Item
Code UNSPSC :
41121804
Matériaux
borosilicate glass
flat bottom flask
short
Joint
joint: ST/NS19/26
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 5
Fabricant/nom de marque
Quickfit FF 50/2S
Disponibilité
not available in USA and Canada
Capacité
50 mL
Description générale
With short neck and ground-glass socket. Not suitable for vacuum use.
Informations légales
Quickfit is a registered trademark of SciLabware Limited