Documents
Z309230
Quickfit® Allihn condenser
L 250 mm, joint: ST/NS 40/38
Synonyme(s) :
CX7/06/SC
About This Item
Code UNSPSC :
41121800
Joint
joint: ST/NS40/38
Fabricant/nom de marque
Quickfit CX 7/06/SC
Disponibilité
not available in USA and Canada
Longueur
250 mm
Superficie
130 cm2
Description générale
Bulb sections give greater surface area for more cooling than the corresponding Liebig type condenser. Generally used for reflux. Condensers have plain stem tops.
Caractéristiques et avantages
• Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)• Also known as reflux or bulb condensers• The inner bulb sections provide an increased cooling surface• Ideal for reflux and Soxhlet extraction applications• Supplied with detachable plastic screw thread connectors which allow 9mm bore flexible tubing to be fitted easily and safely
Informations légales
Quickfit is a registered trademark of SciLabware Limited