Principaux documents
Z301310
Quickfit® plain bends
joint: ST/NS 19/26
Synonyme(s) :
RA9/22
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
joint: ST/NS19/26
Fabricant/nom de marque
Quickfit RA 9/22
Disponibilité
not available in USA and Canada
not available in USA and Canada
Description générale
Socket to cone. Generally used as distilling adapters.
Informations légales
Quickfit is a registered trademark of SciLabware Limited