Principaux documents
Z301124
Quickfit® expansion adapter
top female joint: ST/NS 29/32, bottom male joint: ST/NS 14/23
Synonyme(s) :
XA41
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
female joint: ST/NS top29/32
male joint: ST/NS bottom14/23
Fabricant/nom de marque
Quickfit XA 41
Disponibilité
not available in USA and Canada
not available in USA and Canada
Description générale
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Informations légales
Quickfit is a registered trademark of SciLabware Limited