Principaux documents
Z288365
Microflex® 63-864 Diamond Grip® powder-free latex gloves
size 8‑8-1/2 L
Synonyme(s) :
disposable glove
1 PKG
24,20 €
10 PKG
221,00 €
24,20 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
24,20 €
10 PKG
221,00 €
About This Item
Code UNSPSC :
46181504
24,20 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
latex
natural gloves
Taille
8‑8-1/2 L
Stérilité
non-sterile
non-sterile
Caractéristiques
disposable
disposable
latex free: no
powder-free
Conditionnement
pack of 100 ea (boxes of 100)
pack of 100
Fabricant/nom de marque
Microflex 63-864-090
L × épaisseur
9.8 in. (248 mm) × 6.3 mil (0.16 mm)
Description générale
These gloves are thickest at the fingertips for greater barrier protection as well as textured fingers for gripping in wet and dry conditions. In addition, the gloves are powder-free, fully chlorinated and beaded-cuff.
  • 100% powder-free, non-contaminating
  • Controlled manufacturing process for greater product consistency
  • Fingertip textured for enhanced wet and dry gripping
  • Longer and thicker than typical exam gloves for added protection
Thickness:
  • Cuff: 0.13 mm (5.1 mil)
  • Palm: 0.16 mm (6.3 mil)
  • Finger: 0.23 mm (9.1 mil)
  • Length: 248 mm (9.8")
Autres remarques
Microflex Diamond Grip Plus is in the process of re-branding. The new product name will become Microflex 63-864. Changes effect name and packaging only. No change in material specifications.
Informations légales
Diamond Grip is a registered trademark of Microflex Corp.
Microflex is a registered trademark of CIT Healthcare LLC