Principaux documents
Z286400
FEP sample tube liner
Wilmad 6005-8, O.D. 5 mm
Synonyme(s) :
Wilmad nmr tubes, nmr sample tubes, nmr tubes
About This Item
Code UNSPSC :
41121705
Nomenclature NACRES :
SB.32
Description
Recommended outer tube 528-PP-8
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Wilmad 6005-8
D.E.
5 mm
Volume
80 μL
Description générale
Thin-wall FEP liner minimizes filling factor losses of using inserts and protects NMR tube from etching effects of strong acids containing fluoride ions. Liners are 8 in. L and fit snugly into most common 7 in. L thin-wall tubes. A PTFE plug to close each liner is included. Not recommended for elevated temperature studies.