Principaux documents
Z282049
Coors alumina pestle
for Z281999
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.35
Matériaux
alumina
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Coorstek 60359
L du pilon
57 mm
Description générale
99.5% alumina. Extremely hard and completely nonabsorbent surface. Order mortar separately.
Informations légales
Coors is a trademark of Coorstek, Inc.