Z272612
Flow control pinch valve
for tubing O.D. 5/16 - 3/8in.
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.79
Produits recommandés
Matériaux
Delrin® body
Description générale
Metered scale on valve body adjusts fluid from zero to full flow without fluid contact with valve. Calibration is accurate and repeatable. Valve mounts easily on existing soft wall tube lines without fittings or cutting.
Informations légales
Delrin is a registered trademark of E. I. du Pont de Nemours and Company