Principaux documents
Z266264
Sigma-Aldrich
Polymer-body combination pH electrodes
BNC fitting
1 EA
156,00 €
156,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
156,00 €
About This Item
Code UNSPSC :
41115603
Nomenclature NACRES :
NB.79
156,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Tige, L × diam.
110 mm × 12 mm
Description générale
General-purpose, gel-filled
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
The combination of a pH indicating and reference half-cell in an impact-resistant epoxy body. Designed for applications where breakage is a problem.
  • Environmental - field sampling
  • Industrial - process control
  • Educational - core courses
Rugged bulb, fast response in optimal range of 0 to 12 pH. Suggested temp. range: 0 to 80 °C. For Tris, sulfides, and heavy metals, choose a double junction reference electrode. Supplied with 36 in. lead.