Principaux documents
Z187941
Sigma-Aldrich
Precisionware bottles
Narrow-mouth, capacity 1,000 mL
Synonyme(s) :
bottle hdpe, plastic bottle, plastic bottles
6 EA
54,10 €
24 EA
208,00 €
54,10 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
6 EA
54,10 €
24 EA
208,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
54,10 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
high-density polyethylene bottle
polypropylene cap
round translucent bottle
Joint
Joints threaded neck (38)
Caractéristiques
with cap
Conditionnement
case of 24 ea
pack of 6 ea
Fabricant/nom de marque
Bel-Art F10620-0008
Capacité
1,000 mL
Description générale
  • Precisionware bottles are made of laboratory/pharmaceutical grade high-density polyethylene (HDPE).
  • Features a leak-proof closure cap for sample protection.
  • The sturdy, heavy-duty bottle design facilitates the easy storing of liquid/powder samples.
  • Manufactured to strict DOT-2E specifications.
  • Molded pour lip for easy, no-spill pour.
  • Suitable for shipping.
Bottle with leakproof and sturdy, heavy-duty wall. Manufactured to strict DOT-2E specifications. Molded pour lip for easy, no-spill pour. Suitable for shipping.
Informations légales
Precisionware is a trademark of Vectra Precisionware