Z177849
Scienceware® Ellipso-Spoon® sampler
L 15 cm
Synonyme(s) :
curved laboratory spatula, laboratory spoon
Fabricant/nom de marque
Bel-Art H368060015
Longueur
15 cm
Description générale
Special curved shape permits contact with all corners of vessels, bottles, or containers. No corner out of reach and no loss of materials. Handles crystals, powders, granulated and solid substances, even thick liquids or delicate substances. SS, will not bend. Has both spoon end and spatula end.
Informations légales
Ellipso-Spoon is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP