Principaux documents
Z164062
Sigma-Aldrich
Aldrich® antisplash splash-guard adapter with return hole
female joint: ST/NS 24/40, male joint: ST/NS 24/40, capacity 250 mL
1 EA
159,00 €
159,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
159,00 €
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
159,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Joint
female joint: ST/NS24/40
male joint: ST/NS24/40
Capacité
250 mL
Catégories apparentées
Description générale
These traps greatly reduce carry-over from foaming or bumping. Anything that may climb can return back down through the return hole at the bottom of this adapter.
Application
Aldrich antisplash splash-guard adapter with return hole may be used between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into the condenser in case of "bumping". A drain hole at the base of the inner tube allows solvent to drain back into the evaporation flask.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC